missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS2 Monoclonal Antibody (OTI4B11), TrueMAB™, OriGene
Description
OAS2 Monoclonal antibody specifically detects OAS2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | OAS2 |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI4B11 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gen | OAS2 |
| Gen-Zugriffsnummer | P29728 |
| Gen-Alias | (2-5')oligo(A) synthase 2; (2'-5')oligo(A) synthetase 2; 2',5'-oligoadenylate synthetase-like 11; 2,5-oligoadenylate synthetase-like 11; 2'-5' oligoadenylate synthetase 2; 2'-5' oligoadenylate synthetase-like 11; 2-5A synthase 2; 2'-5'-oligoadenylate synthase 2; 2'-5'-oligoadenylate synthetase 2; 2'-5'-oligoadenylate synthetase 2, 69/71kDa; A synthetase 2; MGC78578; Oas2; Oasl11; p69 OAS / p71 OAS; p69OAS / p71OAS |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?