Learn More
Invitrogen™ Ocrelizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA559573
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ocrelizumab is a CD20-directed cytolytic antibody indicated for the treatment of patients with relapsing or primary progressive forms of multiple sclerosis. It is a second-generation recombinant humanized monoclonal IgG1 antibody that selectively targets the B lymphocytes that express the CD20 antigen. As a humanized molecule, ocrelizumab is expected to be less immunogenic with repeated infusions which improves the benefit-to-risk profile for patients with relapsing or progressive forms of MS.
Spezifikation
| Ocrelizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Ocrevus | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |