missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ODF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA551649
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: HKRGMKGDTV NVRRSVRVKT KNPPHCLEIT PPSSEKLVSV MRLSDLSTED DDSGHCKMNR YDKKIDSLMN AVGCLKSEVK MQKGERQMAK RFLEERKEEL EEVAHELAET EHENTVLRHN IERMKEEKDF TILQKKHLQQ EKE Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Seems to be a major component of sperm tail outer dense fibers (ODF). ODFs are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail and may help to maintain the passive elastic structures and elastic recoil of the sperm tail. May have a modulating influence on sperm motility. Functions as a general scaffold protein that is specifically localized at the distal/subdistal appendages of mother centrioles. Component of the centrosome matrix required for the localization of PLK1 and NIN to the centrosomes. Required for the formation and/or maintenance of normal CETN1 assembly.
Spezifikation
| ODF2 | |
| Polyclonal | |
| Unconjugated | |
| ODF2 | |
| 84 kDa outer dense fiber protein; AI848335; cancer/testis antigen 134; cenexin; cenexin 1; CT134; KKT4; MMTEST29; ODF2; ODF2/1; ODF2/2; Odf84; outer dense fiber of sperm tail 2; outer dense fiber of sperm tails 2; outer dense fiber of sperm tails protein 2; outer dense fiber of sperm tails, 84-kD; outer dense fiber protein 2; RP11-187B3.1; sperm outer dense fiber major protein 2; sperm tail structural protein; testis tissue sperm-binding protein Li 51e; testis-specific autoantigen | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4957 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5BJF6 | |
| ODF2 | |
| Recombinant protein corresponding to Human ODF2. Recombinant protein control fragment (Product #RP-101019). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction