missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ODF2L Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA555766
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: SVKVVHERLQ IQIHKREAEN DKLKEYVKSL ETKIAKWNLQ SRMNKNEAIV MKEASRQKTV ALKKASKVYK QRLDHFTGAI E Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 74%.
ODF2L, also named as KIAA1229, has many isoforms with MW 60-80 kDa. The function of ODF2L (Outer dense fiber protein 2-like) remains largely unknown.
Spezifikation
| ODF2L | |
| Polyclonal | |
| Unconjugated | |
| Odf2l | |
| 4733401D09Rik; 9630045K08Rik; Basal body centriole-associated protein; Bcap; D3Ertd250e; dJ977L11.1; KIAA1229; ODF2L; outer dense fiber of sperm tails 2 like; outer dense fiber of sperm tails 2-like; outer dense fiber protein 2-like; Protein BCAP; RGD1308397 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57489 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULJ1 | |
| Odf2l | |
| Recombinant protein corresponding to Human ODF2L. Recombinant protein control fragment (Product #RP-94165). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur