missing translation for 'onlineSavingsMsg'
Learn More

Oligomycin, TRC

Product Code. 30428451
Change view
Click to view available options
Menge:
5 mg
Unit Size:
5 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30428451 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428451 Supplier Toronto Research Chemicals Supplier No. TRCO5329735MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Oligomycin
CAS 1404-19-9
Summenformel C45H74O11
Menge 5 mg
Synonym Oligomycin A','B','C mixture
InChI-Formel InChI=1S/C45H74O11/c1-12-34-17-15-13-14-16-27(4)42(51)44(11,53)43(52)32(9)40(50)31(8)39(49)30(7)38(48)26(3)18-21-37(47)54-41-29(6)35(20-19-34)55-45(33(41)10)23-22-25(2)36(56-45)24-28(5)46/h13-15,17-18,21,25-36,38,40-42,46,48,50-51,53H,12,16,19-20,22-24H2,1-11H3/b14-13+,17-15+,21-18+/t25-,26-,27+,28+,29+,30-,31-,32-,33-,34-,35-,36-,38+,40+,41+,42-,44+,45-/m1/s1
SMILES CC[C@](CC[C@]1([H])[C@H](C)[C@]([C@@H](C)[C@]2(CC[C@@H](C)[C@](C[C@@H](O)C)([H])O2)O1)([H])OC3=O)([H])/C=C/C=C/C[C@H](C)[C@@H](O)[C@@](O)(C)C([C@H](C)[C@@H](O)[C@H](C)C([C@H](C)[C@@H](O)[C@H](C)/C=C/3)=O)=O
IUPAC-Name (1S,4E,5'R,6R,6'R,7S,8R,10S,11S,12R,14S,15R,16S,18E,20E,22S,25R,27S,28R,29S)-22-Ethyl-7,11,14,15-tetrahydroxy-6'-((S)-2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-3',4',5',6'-tetrahydro-2,26-dioxaspiro[bicyclo[23.3.1]nonacosan-27,2'-pyran]-4,18,20-trien-3,9,13-trion
Molekulargewicht (g/mol) 791.06
Formelmasse 790.52
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.