missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Omipalisib

Click to view available options
Menge:
10 mg
50 mg
Unit Size:
10 Milligramm
50 Milligramm
Description
Omipalisib is a highly potent PI 3-K inhibitor (IC50 = 0.04 nM). Also inhibits mTOR (Ki = 0.18 - 0.3 nM) and DNA-PK (IC50 = 0.28 nM). Induces G1 cell cycle arrest and inhibits proliferation of some breast cancer cells lines in vitro. Reduces pAKT-S473 levels in and inhibits growth of BT474 breast cancer xenografts in mice. Orally bioavailable.
Specifications
Specifications
| Chemischer Name oder Material | 2,4-Difluoro-N-[2-methoxy-5-[4-(4-pyridazinyl)-6-quinolinyl]-3-pyridinyl]benzenesulfonamide |
| Synonym | GSK2126458 |
| Menge | 10 mg |
| Summenformel | C25H17F2N5O3S |
| Formelmasse | 505.5 |
| Produkttyp | Hemmstoff |
| Reinheit | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction