Learn More
OPN, SPP1 Rabbit, anti-Human, Clone: 6B11, Proteintech
Rabbit Recombinant Monoclonal Antibody
Marke: Proteintech 80912-4-RR-20UL
Beschreibung
The OPN, SPP1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human OPN, SPP1. This antibody recognizes human, rat, mouse antigen. The OPN, SPP1 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| OPN, SPP1 | |
| 6B11 | |
| BNSP, Bone sialoprotein 1, BSPI, ETA 1, Nephropontin, OPN, Osteopontin, secreted phosphoprotein 1, SPP 1, SPP1, Urinary stone protein, Uropontin | |
| 20 μL | |
| 6696 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse | |
| 150 kDa | |
| IgG |