missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OPRM1 Monoclonal Antibody (677014)
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561732
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
The OPRM1 gene encodes the mu opioid receptor, which is the primary site of action for the most commonly used opioids, including morphine, heroin, fentanyl, and methadone. It is also the primary receptor for endogenous opioid peptides beta-endorphin and the enkephalins. The OPRM1 receptor is a membrane of the G protein-coupled receptor family. There are at least 3 types of opioid receptors, mu, kappa, and delta, each with a distinct pharmacologic profile.
Spezifikation
| OPRM1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| P33535 | |
| OPRM1 | |
| Synthetic peptide corresponding to His385-Pro398 of rat μ Opioid R/OPRM1. | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 677014 | |
| Unconjugated | |
| OPRM1 | |
| hMOP; KIAA0403; LMOR; MOP; MOP receptor; MOP-R; Mor; MOR1; M-OR-1; MOR-1; MOR-1C; MOR-1O; MORA; Mu opiate receptor; mu opioid receptor; mu opioid receptor hMOR-1a; mu opioid receptor splice variant mMOR-1Z; mu opioid receptor splice variant rMOR-1S; mu opioid receptor splice variant rMOR-1Z; mu-opioid receptor; muOR; MUOR1; mu-type opioid receptor; Opioid mu receptor; Opioid Receptor; opioid receptor B; opioid receptor mu 1; opioid receptor, mu 1; OPRM; Oprm1; Oprrm1; OTTHUMP00000017463; Ror-b | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 25601 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur