missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Opucolimab Recombinant Monoclonal Antibody
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Opucolimab, also known as HLX20, is a recombinant human monoclonal antibody that targets the programmed death-ligand 1 (PD-L1), also referred to as CD274 or B7-H1. CD274 is a gene encoding a protein critical for immune checkpoint regulation. Located on chromosome 9, this protein is expressed on the surface of various cell types, including tumor cells, immune cells, and normal tissues. PD-L1 interacts with the PD-1 receptor on T cells to modulate immune responses, particularly by promoting self-tolerance and preventing autoimmunity. In the context of cancer, aberrant expression of CD274 allows tumor cells to evade immune surveillance, leading to immune escape. This mechanism has made CD274 a significant target for cancer immunotherapies, particularly immune checkpoint inhibitors such as pembrolizumab and nivolumab, which block the PD-1/PD-L1 interaction to restore anti-tumor immunity. Research on CD274 continues to reveal its role in various malignancies and its potential as a prognostic and therapeutic biomarker.
Spezifikation
Spezifikation
| Antigen | Opucolimab Humanized |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Klassifikation | Recombinant Monoclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Wirtsspezies | Human |
| Reinigungsverfahren | Protein A |
| Menge | 1 mg |
| Regulatorischer Status | RUO |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?