Learn More
Edmund Buhler KREISSCHUETTLER ROTATEST 560
Click to view available options
Drehzahlbereich:
15 bis 300 U/min
Packungsgröße:
1 Stück
Beschreibung
- Crosschuettler Rotatest 560
- Speed 15 to 300 rpm; Amplitude 26 mm; Antiskid plate 56 x 40 cm; Maximum load 30 kg; Shaker plate 400; Max. Load 15 Kg
- Platform agitators 2 to 30kg
- Reciprocating movement, orbital, reciprocating and orbital combination (quick choice by lever action) depending on model
- Platform with 3 fixing rollers and non-slip rubber mat included for 10449882
- Complete with ref 10736871 platform 5 fixing rollers and non-slip rubber mat included for 11834251, 11844251, 10059380
- Operating condition 5-50 ° C, 85% Hr
Spezifikation
Spezifikation
| Tragkraft (metrisch) | 30 kg |
| Drehzahlbereich | 15 bis 300 U/min |
| Produkttyp | Orbitalschüttler |
| Versandgewicht (metrisch) | 32 kg |
| Modell | Rotatest 560 |