missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteopontin/OPN Antibody (CL10686) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-07980V
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Osteopontin/OPN Monoclonal antibody specifically detects Osteopontin/OPN in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Spécification
| Osteopontin/OPN | |
| Monoclonal | |
| DyLight 405 | |
| BNSP, Bone sialoprotein 1, MGC110940, Nephropontin, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
| This antibody was developed using a recombinant protein derived from P10451, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Extracellular Matrix, Hypoxia | |
| 6696 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10686 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu