Learn More
Invitrogen™ Otelixizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559659
 View more versions of this product
Product Code. 30283135
  • 477.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
This item is not returnable. View return policy
Description
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Otelixizumab is a monoclonal antibody that binds to a receptor found on all T cells called CD3, which is involved in normal T cell signaling. Otelixizumab is believed to inhibit the function of autoreactive T cells, which are important in propagating autoimmune diseases, while inducing regulatory T cell pathways that promote immunological tolerance and inhibit autoreactive disease activity. Tolerx is developing otelixizumab to treat patients with type 1 diabetes, psoriasis and other autoimmune diseases such as rheumatoid arthritis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Otelixizumab Humanized
Recombinant Monoclonal
Unconjugated
ChAglyCD
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications