missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ovalbumin Polyclonal Antibody, HRP

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA1196HRP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
By Western blot, PA1-196-HRP detects ovalbumin at ~45kD. Multiple bands may be observed, which are likely due to the presence of multiple glycosylated and/or aggregated species of ovalbumin within a particular protein preparation.
Chicken ovalbumin is the major protein in the 'white' of the egg (and a favorite antigen in immunological research). Egg white contains a variety of proteins including ovalbumin, conalbumin, ovomucoi and lysozyme. It belonds to the serpin family and the Ov serpin subfamily. Ovalbumin can cause an allergic reaction in humans. Ovalbumin has been implicated in the development of the egg shell. Immunohistochemistry revealed that ovalbumin is found only in the mammillary bodies of decalcified shell, and is not distributed throughout the shell matrix. These results indicate that ovalbumin is present during the initial phase of shell formation and becomes incorporated into the protein matrix of the mammillary bodies. However, it is not yet clear whether ovalbumin at this site plays a specific role in shell mineralization.
Spezifikation
| Ovalbumin | |
| Polyclonal | |
| HRP | |
| OVAL | |
| Allergen Gal d II; clade (ovalbumin); egg albumin; Gal d 2; member 14; OVA; OVAL; Ovalbumin; ovalbumin (SERPINB14); plakalbumin; serpin peptidase inhibitor; SERPINB14; unnamed protein product | |
| Rabbit | |
| Protein G | |
| RUO | |
| 396058 | |
| 4°C | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with proprietary stabilizer and 0.07% Kathon | |
| P01012 | |
| OVAL | |
| Ovalbumin. | |
| 100 μg | |
| Primary | |
| Chicken | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur