missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ozuriftamab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA560072
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ozuriftamab is a biosimilar targeting the receptor tyrosine kinase orphan receptor 2 (ROR2). The ROR2 gene encodes the protein receptor tyrosine kinase-like orphan receptor 2 (ROR2), which is involved in non-canonical Wnt signaling pathways. ROR2 is known to play critical roles in skeletal development, particularly in the formation of limb bones and cartilage. The gene is located on chromosome 9 and consists of 9 exons that encode a protein of 943 amino acids with a single-pass transmembrane receptor structure. ROR2's extracellular region contains an immunoglobulin-like domain, a cysteine-rich domain (CRD), and a kringle domain, which facilitate its interaction with Wnt ligands such as Wnt5a and Wnt11. Upon ligand binding, ROR2 can homodimerize to initiate downstream signaling cascades that influence cell polarity, migration, and differentiation. Aberrations in ROR2 expression and function are implicated in various disorders, including Robinow syndrome and recessive renal cancer. Additionally, ROR2 has been noted to play a role in cancer cell motility and invasion, highlighting its potential as a therapeutic target.
Spezifikation
| Ozuriftamab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur