missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-selectin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Spezifikation
| Antigen | P-selectin |
|---|---|
| Konzentration | 0.25 mg/mL |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16861444
|
Proteintech
13304-1-AP-20UL |
20 μL |
145.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16851444
|
Proteintech
13304-1-AP-150UL |
150 μL |
410.00 €
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
P-Selectin, also called CD62, GRMP, GMP140 is a 140-kD adhesion molecule, expressed at the surface of activated cells, that mediates the interaction of activated endothelial cells or platelets with leukocytes. The P-Selectin gene is mapped on 1q24.2. P-selectin plays an essential role in the initial recruitment of leukocytes (white blood cells) to the site of injury during inflammation. P-selectin is also very important in the recruitment and aggregation of platelets at areas of vascular injury. In quiescent platelet, P-selectin is located on the inner wall of alpha-granules. Sorting nexin-17 interacts with the cytosolic domain of P-selectin and suggested that SNX17 may function in the intracellular trafficking of P-selectin. P-selectin expression was found to be significantly and positively correlated with carotid artery stiffness and wall thickness, and the percentage of P-selectin-positive platelets was significantly higher in patients with carotid plaque.Spezifikation
| P-selectin | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P16109 | |
| 6403 | |
| P-selectin Fusion Protein Ag4124 | |
| Primary | |
| -20°C | |
| SELP |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CD62, CD62P, GMP 140, GMP140, GMRP, Granule membrane protein 140, GRMP, LECAM3, P selectin, PADGEM, PSEL, P-selectin, SELP | |
| SELP | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts