Learn More
P21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 10355-1-AP-20UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16846693
  • 145.00 € / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Protein p21 is a tumor suppressor protein which is an inhibitor of Cyclin-dependent kinases (CDK) and is transcriptionally activated by p53. P21 is important in the response of cells to genotoxic stress and a major transcriptional target of p53 protein. The occurrence of p21 in the nucleus executes binding and inhibition activity of cyclin dependent kinases Cdk1 and Cdk2, and blocks the transition from G1 phase into S phase or from G2 phase into mitosis after DNA damage, which enables the repair of damaged DNA. In the cytoplasm, p21 protein has an anti-apoptotic effect. P21 is able to bind to and inhibit caspase 3, as well as the apoptotic kinases ASK1 and JNK. P21 exhibits a dual function in carcinogenesis, and acts as a tumor suppressor, prevents apoptosis, and acts as an oncogene.
Spezifikation
Spezifikation
P21
Polyclonal
Unconjugated
CDKN1A
CAP20, CDK interacting protein 1, CDKN1, CDKN1A, CIP1, MDA 6, MDA6, P21, p21CIP1, PIC1, SDI1, WAF1
Rabbit
Antigen Affinity Chromatography
RUO
1026
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunofluorescence
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P38936
CDKN1A
P21 Fusion Protein Ag0368
20 μL
Primary
Human
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen