missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p38 MAPK gamma Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA5148517
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
MAPK12 (p38 gamma) is a serine/threonine kinase involved in many different cellular processes. It is activated during hypoxia and gamma-irradiation-induced G(2) arrest. Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. MAPK12 functions as a signal transducer during differentiation of myoblasts to myotubes.
Spezifikation
| p38 MAPK gamma | |
| Polyclonal | |
| Unconjugated | |
| Mapk12 | |
| AW123708; ERK3; ERK5; erk6; ERK-6; etID309866.18; Extracellular signal-regulated kinase 6; fa05c12; LOW QUALITY PROTEIN: mitogen-activated protein kinase 12; mitogen-activated protein kinase 12; MAP kinase 12; MAP kinase p38 gamma; MAPK 12; mapk12; mapk12a; mitogen activated protein kinase 12; mitogen-activated protein kinase 12; mitogen-activated protein kinase 12a; mitogen-activated protein kinase 3; mitogen-activated protein kinase p38 gamma; OTTHUMP00000196671; P38gamma; p38-gamma; Prkm12; SAP kinase-3; sapk3; SAPK-3; stress activated protein kinase 3; stress-activated protein kinase 3; stress-activated protein kinase-3; wu:fa05c12; zgc:101695 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 29857, 60352, 6300 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O08911, P53778, Q63538 | |
| Mapk12 | |
| E. coli-derived recombinant human p38 gamma , Met1-Leu367. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction