missing translation for 'onlineSavingsMsg'
Learn More

p53 Antibody (TP53/3890R), FITC, R&D Systems™

Product Code. 18612394 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
18612394 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18612394 Supplier Novus Biologicals Supplier No. NBP321044F
 Eingeschränkter Artikel
Verfügbar ab: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Anwendungen Flow Cytometry, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon TP53/3890R
Konjugat FITC
Zusammensetzung PBS
Gen-Alias Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
Wirtsspezies Rabbit
Immunogen Recombinant human full-length p53 protein
Reinigungsverfahren Protein A or G purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primär oder sekundär Primary
Gen-ID (Entrez) 7157
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Produkttyp Antibody
Form Purified
Isotype IgG κ
Produktlinie Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.