missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ p73 Recombinant Rabbit Monoclonal Antibody (K01_3K31)

Artikelnummer. 30829294
Ansicht ändern
Click to view available options
Menge:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30829294 100 μL
30828117 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30829294 Lieferant Invitrogen™ Lieferanten-Nr. MA564345
 Eingeschränkter Artikel
Verfügbar ab: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

p73 Recombinant Monoclonal Antibody for Western Blot

P73 (TP73) participates in the apoptotic response to DNA damage. Isoforms containing the transactivation domain are pro-apoptotic, isoforms lacking the domain are anti-apoptotic and block the function of p53 and transactivating p73 isoforms. May be a tumor suppressor protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p73
Anwendungen Western Blot
Klassifikation Recombinant Monoclonal
Klon K01_3K31
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gen TP73
Gen-Zugriffsnummer O15350, Q9JJP2
Gen-Alias deltaNp73; OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; P73; RP23-254N4.10; TAp73; Tp73; transformation related protein 73; Trp73; Tumor protein p73
Gensymbole TP73, Trp73
Wirtsspezies Rabbit
Immunogen Peptide.
Reinigungsverfahren Protein A/G
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 22062, 7161
Zielspezies Human, Mouse
Inhalt und Lagerung Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.