missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladin Antibody (1E6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 16 publications
Marke: Novus Biologicals NBP1-25959SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Palladin Monoclonal specifically detects Palladin in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.
Spezifikation
| Palladin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, Flow Cytometry 5 ug/ml, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 0.05 ml 2 ml lysate, Immunohistochemistry-Paraffin 1:50-1:100 | |
| CGI151, CGI-151, FLJ22190, FLJ38193, KIAA0992FLJ39139, MYN, myoneurin, palladin, palladin, cytoskeletal associated protein, PNCA1, Sarcoma antigen NY-SAR-77, SIH002FLJ61376 | |
| Mouse | |
| 85 kDa | |
| 0.025 mL | |
| Cellular Markers, Cytoskeleton Markers, Signal Transduction | |
| 23022 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1000000 | |
| Unconjugated | |
| Q8WX93 | |
| PALLD | |
| Endogenous Palladin immunoprecipitated from chicken embryo fibroblasts. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur