Learn More
Invitrogen™ Palladin Recombinant Rabbit Monoclonal Antibody (24GB4125)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557151
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Palladin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin's different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.
Spezifikation
| Palladin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PALLD | |
| 2410003B16Rik; 6030492A02; 90kDa actin-associated protein palladin; actin-associated protein palladin; CGI151; CGI-151; FLJ22190; FLJ38193; FLJ39139; involved in cytoskeletal assembly; KIAA0992; MYN; myoneurin; palladin; palladin isoform; palladin pseudogene; palladin, cytoskeletal associated protein; PALLD; PNCA1; Sarcoma antigen NY-SAR-77; SIH002 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23022, 364558, 72333 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4125 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P0C5E3, Q8WX93, Q9ET54 | |
| PALLD | |
| A synthesized peptide derived from human Palladin (aa 1-65). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |