Learn More
Pan-PAX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21383-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody is a Pan-PAX antibody.
This gene encodes a member of the paired box (PAX) family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternatively spliced transcript variants encoding different isoforms have been described.Spezifikation
| Pan-PAX | |
| Polyclonal | |
| Unconjugated | |
| PAX1 | |
| paired box, paired box 8, Paired box protein Pax, Paired box protein Pax 8, Pan-PAX, PAX 8, PAX8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114502, 18503, 18504, 18505, 18506, 18507, 18508, 18509, 18510, 18511, 25509, 293992, 311505, 362741, 500453, 500574, 5075, 5076, 5077, 5078, 5079, 5080, 5081, 5083, 7849, 81819, 83630 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43316, O88436, P09084, P15863, P23759, P23760, P24610, P26367, P32114, P32115, P47239, P47242, P51974, P55771, P63015, P63016, Q00288, Q02548, Q02650, Q02962, Q06710, Q2L4T2 | |
| Pax1, Pax2, PAX3, PAX4, PAX5, PAX6, PAX7, Pax8, PAX9 | |
| Pan-PAX Fusion Protein Ag16018 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |