missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pancreatic Amylase Monoclonal Antibody (AMT10-105.8)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA110619
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MA1-10619 detects Amylase from human samples. MA1-10619 has been successfully used in ELISA applications. The MA1-10619 immunogen is purified human pancreatic Amylase.
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch andglycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas.
Spezifikation
| Pancreatic Amylase | |
| Monoclonal | |
| 2.2 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P04746, P19961 | |
| AMY2A, AMY2B | |
| Purified human pancreatic Amylase. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| ELISA | |
| AMT10-105.8 | |
| Unconjugated | |
| AMY2B | |
| 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a4; AMY2B; AMY3; amylase; amylase 2-2, pancreatic; amylase 2a1; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; ENSMUSG00000074267; glycogenase; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A | |
| Mouse | |
| Purified | |
| RUO | |
| 279, 280 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur