Learn More
Thermo Scientific Chemicals Pancreatin, porcine pancreas
Description
Hydrolyzes starch into dextrins and sugar Pancreatin from porcine pancreas is used in the treatment of steatorrhea by lipase supplementation therapy and for pancreatic diabetes. It is also used in the removal of formalin-fixed tissues from arterial grafts without causing damage in fiber integrity. Further, it finds uses in contact lenses. In addition to this, it hydrolyzes starch into dextrins and sugar. It is also used to treat conditions responsible for deficiency in pancreatic secretions, such as surgical pancreatectomy, pancreatitis and cystic fibrosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Menge | 1kg |
| Verpackung | Glass bottle |
| Form | Lyophilized |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.