Learn More
Thermo Scientific Chemicals Pancreatin, porcine pancreas
Artikelnummer. 15414589
Change view
Click to view available options
Menge:
1kg
Packungsgröße:
1 Kilogramm
Artikelnummer. Menge unitSize
15414589 1kg 1 Kilogramm
1 options
Artikelnummer. 15414589
Marke: Thermo Scientific Chemicals J62162.A1
Bulk oder benutzerdefiniertes Format anfordern
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen
Hydrolyzes starch into dextrins and sugar Pancreatin from porcine pancreas is used in the treatment of steatorrhea by lipase supplementation therapy and for pancreatic diabetes. It is also used in the removal of formalin-fixed tissues from arterial grafts without causing damage in fiber integrity.
Hydrolyzes starch into dextrins and sugar Pancreatin from porcine pancreas is used in the treatment of steatorrhea by lipase supplementation therapy and for pancreatic diabetes. It is also used in the removal of formalin-fixed tissues from arterial grafts without causing damage in fiber integrity. Further, it finds uses in contact lenses. In addition to this, it hydrolyzes starch into dextrins and sugar. It is also used to treat conditions responsible for deficiency in pancreatic secretions, such as surgical pancreatectomy, pancreatitis and cystic fibrosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Spezifikation
Menge 1kg
Verpackung Glass bottle
Form Lyophilized