missing translation for 'onlineSavingsMsg'
Learn More

Panobinostat, MedChemExpress

Product Code. 16493718
Encompass_Preferred
Change view
Click to view available options
Menge:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
10 Milligramm
100 Milligramm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16493718 10 mg 10 Milligramm
16450956 200 mg -
16450107 50 mg -
16424796 100 mg 100 Milligramm
16471546 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493718 Supplier Medchem Express Supplier No. HY10224

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

MedChemExpress Panobinostat (LBH589; NVP-LBH589) is a potent and orally active non-selective HDAC inhibitor, and has antineoplastic activities. Panobinostat induces HIV-1 virus production even at low concentration range 8-31 nM, stimulates HIV-1 expression in latently infected cells. Panobinostat induces cell apoptosis and autophagy. Panobinostat can be used for the study of refractory or relapsed multiple myeloma.

Panobinostat (LBH589

Specifications

Chemischer Name oder Material Panobinostat
Physikalische Form Powder
CAS 404950-80-7
Farbe Light Yellow
Menge 10 mg
Hinweise zur Reinheitsqualität Research
Summenformel C21H23N3O2
Zur Verwendung mit (Anwendung) Cancer-programmed cell death
Synonym LBH589 NVP-LBH589
Löslichkeitsinformationen DMSO : ≥ 57 mg/mL (163.12 mM)
SMILES O=C(/C=C/C1=CC=C(CNCCC2=C(NC3=C2C=CC=C3)C)C=C1)NO
Molekulargewicht (g/mol) 349.43
Formelmasse 349.43
Reinheit (%) 95.33%
Güte Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.