missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax7 Antibody (PAX7/497), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-34706H
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Pax7 Monoclonal specifically detects Pax7 in Human, Mouse, Rat, Chicken, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Spezifikation
| Pax7 | |
| Monoclonal | |
| HRP | |
| FLJ37460, HuP1, paired box 7, paired box gene 7, paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7, PAX7 transcriptional factor, PAX7B, RMS2 | |
| Mouse | |
| 57 kDa | |
| 0.1 mL | |
| Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 5081 | |
| Human, Mouse, Rat, Chicken, Zebrafish | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry (Frozen) | |
| PAX7/497 | |
| Western Blot, ELISA, Immunohistochemistry-Frozen | |
| PAX7 | |
| Recombinant fragment (aa301-505) of human Pax7 protein (exact sequence is proprietary) (Uniprot: P23759) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| The Pax gene family of nuclear transcription factors is comprised of nine members that function during embryogenesis to regulate the temporal and position-dependent differentiation of cells. In addition, the family is involved in a variety of signal transduction pathways in the adult organism. Mutations in the Pax family of proteins have been linked to disease and cancer in humans. Pax-7 is a protein specifically expressed in cultured satellite cell-derived myoblasts. In situ hybridization reveals that Pax-7 is also expressed in satellite cells residing in adult muscle. A chromosomal aberration in the gene encoding Pax-7 causes rhabdomyosarcoma 2 (RMS2) (also called alveolar rhabdomyosarcoma). | |
| Store at 4C in the dark. | |
| IgG1 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur