missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHGC5 Mouse anti-Human, Clone: OTI2G2, lyophilized, TrueMAB™
Beschreibung
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Spezifikation
Spezifikation
| Antigen | PCDHGC5 |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI2G2 |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 8% trehalose and no preservative; pH 7.3 |
| Gen | PCDHGC5 |
| Gen-Zugriffsnummer | Q9Y5F6 |
| Gen-Alias | PCDH-GAMMA-C5 |
| Gensymbole | PCDHGC5 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?