missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCK1 Polyclonal Antibody

Sheep Polyclonal Antibody
Marke: Invitrogen™ PA5149250
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
Phosphoenolpyruvate Carboxykinase 1 (PCK1) is a main control point for the regulation of gluconeogenesis. During gluconeogenesis PCK1 acts as the rate-limiting enzyme. PCK1 regulates the formation and maintenance of memory CH8(+) T-cells via gluconeogenesis. PCK1 aids in controlling the levels of metabolic intermediates in the citric acid cycle by regulating cataplerosis and anaplerosis. When glucose levels are high, PCK1 catalyzes the cataplerotic conversion of oxaloacetate to phosphoenolpyruvate (PEP). When they are low it catalyzes the anaplerotic conversion of phosphoenolpyruvate to oxaloacetate. When phosphorylated at Ser-90 by AKT1, PCK1 acts as a protein kinase by reducing the binding affinity to oxaloacetate and promotes an atypical serine protein kinase activity using GTP as the donor. This activity regulates lipogenesis by disrupting the interaction between INSIG proteins and SCAP and promoting nuclear translocation of SREBP proteins and transcription of lipogenesis-related genes. An important paralog of this gene is PCK2.
Spezifikation
| PCK1 | |
| Polyclonal | |
| Unconjugated | |
| PCK1 | |
| AI265463; GTP; HGNC:8724; MGC22652; PCK; Pck1; Pck-1; PE PEPCKC; PEP carboxykinase; PEPCK; PEPCK1; PEPCKC; PEPCK-C; phosphoenolpyruvate carboxykinase (GTP); phosphoenolpyruvate carboxykinase 1; phosphoenolpyruvate carboxykinase 1 (soluble); phosphoenolpyruvate carboxykinase 1, cytosolic; phosphoenolpyruvate carboxykinase, cytosolic; phosphoenolpyruvate carboxykinase, cytosolic (GTP); phosphoenolpyruvate carboxykinase, cytosolic [GTP]; phosphoenolpyruvate carboxylase; phosphopyruvate carboxylase; RATPEPCK | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 18534, 362282, 5105 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P07379, P35558, Q9Z2V4 | |
| PCK1 | |
| E. coli-derived recombinant mouse PCK1, Lys551-Met622. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur