missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCM1 Antibody (CL0206) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-52878MFV450
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PCM1 Monoclonal antibody specifically detects PCM1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
| PCM1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| hPCM-1, pericentriolar material 1, pericentriolar material 1 protein, PTC4PCM-1 | |
| This antibody was developed using a recombinant protein derived from Q15154, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Mitotic Regulators | |
| 5108 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0206 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion