Learn More
Applied Biosystems™ PCR Product Pre-Sequencing Kit
The PCR Product Pre-Sequencing Kit uses a novel, enzymatic clean-up method to pre-treat PCR products prior to sequencing, without any subsequent purification or separation steps.
Marke: Applied Biosystems™ 70995.1.KT