Learn More
PCSK9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55206-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody is specific to PCSK9.
This gene encodes a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. The encoded protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. This protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons. Mutations in this gene have been associated with a third form of autosomal domit familial hypercholesterolemia.Spezifikation
| PCSK9 | |
| Polyclonal | |
| Unconjugated | |
| PCSK9 | |
| FH3, HCHOLA3, LDLCQ1, NARC 1, NARC1, PC9, PCSK9, Proprotein convertase 9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100102, 255738, 298296 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59996, Q80W65, Q8NBP7 | |
| Pcsk9 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |