missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PD-L1/VEGF Biosimilar Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Artikelnummer. 30284642
Change view
Click to view available options
Menge:
1 mg
100 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
Artikelnummer. Menge unitSize
30284642 100 μg 100 Mikrogramm
30282815 1 mg 1 Milligramm
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30284642

Marke: Invitrogen™ MA560151

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Human Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Pm8002.

The PD-L1 (Programmed Death-Ligand 1) gene, located on chromosome 9p24.1, is essential for immune response modulation and evasion by cancer cells. It encodes a protein comprising extracellular IgV and IgC domains, a transmembrane domain, and a short cytoplasmic tail. PD-L1 binds to the PD-1 receptor on T cells, inhibiting their activity to allow tumor cells to evade immune detection. PD-L1 expression is regulated at multiple levels: genetic, epigenetic, transcriptional, and post-transcriptional. Similarly, VEGF (Vascular Endothelial Growth Factor) is a critical regulator of angiogenesis and vasculogenesis, located on chromosome 6p21.1. VEGF-A, the most studied isoform, promotes endothelial cell proliferation, migration, and survival, primarily through binding to VEGFR1 and VEGFR2 receptors. VEGF is pivotal in both physiological processes like wound healing and pathological conditions such as cancer, where it facilitates tumor growth by enhancing blood supply.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1/VEGF Biosimilar
Anwendungen ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Klassifikation Recombinant Monoclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Wirtsspezies Human
Reinigungsverfahren Protein A
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.