missing translation for 'onlineSavingsMsg'
Learn More

PD-L2/B7-DC/PDCD1LG2 Antibody (S02-2A1), Novus Biologicals™

Artikelnummer. 18641084 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
50 μg
Unit Size:
100 Mikroliter
50 Mikroliter
Product Code. Menge unitSize
18641084 100 μg 100 Mikroliter
18634055 50 μg 50 Mikroliter
2 options
This item is not returnable. View return policy

Product Code. 18641084

Brand: Novus Biologicals NBP319490100ul

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PD-L2/B7-DC/PDCD1LG2 Monoclonal antibody specifically detects PD-L2/B7-DC/PDCD1LG2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Anwendungen Western Blot
Klassifikation Monoclonal
Klon S02-2A1
Konzentration 0.3 mg/mL
Konjugat Unconjugated
Zusammensetzung 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gen-Alias B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Wirtsspezies Rabbit
Immunogen A synthetic peptide of human PD-L2/B7-DC/PDCD1LG2 (Uniprot # Q9BQ51)
Reinigungsverfahren Affinity purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Apoptosis
Primär oder sekundär Primary
Gen-ID (Entrez) 80380
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.