missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDGFRL Monoclonal Antibody (1010639)

Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
PDGFRL bears significant sequence similarity to the ligand binding domain of platelet-derived growth factor receptor beta. Mutations in the PDGFRL gene, or deletion of a chromosomal segment containing this gene, are associated with sporadic hepatocellular carcinomas, colorectal cancers, and non-small cell lung cancers. This suggests PDGFRL may function as a tumor suppressor.
Spezifikation
Spezifikation
| Antigen | PDGFRL |
| Anwendungen | Neutralization |
| Klassifikation | Monoclonal |
| Klon | 1010639 |
| Konzentration | 0.5 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 5% trehalose and no preservative; pH 7.4 |
| Gen | PDGFRL |
| Gen-Zugriffsnummer | Q6PE55 |
| Gen-Alias | 1110039P19Rik; AV013190; PDGF receptor beta-like tumor suppressor; PDGFRL; PDGFR-like protein; PDGRL; platelet derived growth factor receptor like; platelet-derived growth factor receptor-like; platelet-derived growth factor receptor-like protein; platelet-derived growth factor-beta-like tumor suppressor; PRLTS |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?