missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDI Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 16356804
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
Artikelnummer. Menge unitSize
16356804 100 μL 100 Mikroliter
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16356804

Marke: Invitrogen™ PA578098

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse liver Predicted Reactivity: Rat (92%), Rabbit (91%), Chicken (83%), Chimpanzee (100%), Bovine (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Protein disulfide isomerase (PDI) is a multifunctional protein that catalyzes the formation, breakage and rearrangement of disulfide bonds. At the cell surface, the protein seems to act as a reductase that cleaves disulfide bonds of proteins attached to the cell. It may therefore cause structural modifications of exofacial proteins. Inside the cell, PDI seems to form/rearrange disulfide bonds of nascent proteins. At high concentrations, it functions as a chaperone that inhibits aggregation of misfolded proteins. At low concentrations, it facilitates aggregation (anti-chaperone activity). PDI may be involved with other chaperones in the structural modification of the TG precursor in hormone biogenesis. It also acts a structural subunit of various enzymes such as prolyl 4-hydroxylase and microsomal triacylglycerol transfer protein MTTP. Receptor for LGALS9; the interaction retains P4HB at the cell surface of Th2 T helper cells, increasing disulfide reductase activity at the plasma membrane, altering the plasma membrane redox state and enhancing cell migration.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen PDI
Anwendungen Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gen P4hb
Gen-Zugriffsnummer P07237, P09103
Gen-Alias 1810041F13Rik; AI661267; CaO19.12595; cellular thyroid hormone binding protein; cellular thyroid hormone-binding protein; CLCRP1; collagen prolyl 4-hydroxylase beta; DSI; Endoplasmic reticulum resident protein 59; Eps1p; ER protein 59; ERBA2L; ERp59; GIT; glutathione-insulin transhydrogenase; I79_002399; LOW QUALITY PROTEIN: protein disulfide-isomerase A2; MFP1; P4HB; p4hb.L; P4Hbeta; p55; p58; Pancreas-specific protein disulfide isomerase; pancreatic protein disulfide isomerase; PDA2; PDI; PDI (E.C.5.3.4.1); PDI1; PDI1P; Pdia1; pdia2; pdia2.L; PDIP; Pdipl; PDIR; PHDB; PO4DB; PO4HB; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), beta polypeptide; PROHB; prolyl 4-hydroxylase beta polypeptide; Prolyl 4-hydroxylase subunit beta; prolyl 4-hydroxylase subunit beta L homeolog; prolyl 4-hydroxylase subunit beta L homeolog; prolyl 4-hydroxylase, beta polypeptide L homeolog; prolyl 4-hydroxylase, beta polypeptide; prolyl 4-hydroxylase, beta subunit; protein disulfide isomerase; protein disulfide isomerase (pancreas) like; Protein disulfide isomerase (Prolyl 4-hydroxylase, beta polypeptide); protein disulfide isomerase A2; protein disulfide isomerase associated 2; protein disulfide isomerase family A member 2; protein disulfide isomerase family A member 2 L homeolog; protein disulfide isomerase family A, member 1; protein disulfide isomerase family A, member 2; protein disulfide isomerase PDI1; protein disulfide isomerase, pancreatic; protein disulfide isomerase/oxidoreductase; protein disulfide isomerase-associated 1; protein disulfide isomerase-associated 2; protein disulfide-isomerase; Protein disulfide-isomerase A2; protocollagen hydroxylase; Rho GDP dissociation inhibitor gamma; testicular secretory protein Li 32; Thbp; Thioredoxin-related glycoprotein 1; thyroid hormone-binding protein p55; TRG1; XELAEV_18043753mg; XELAEV_18045582mg; XPDIp; YCL043C; YCL313; YCL43C
Gensymbole P4hb
Wirtsspezies Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PDI. The exact sequence is proprietary.
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 18453, 5034
Zielspezies Human, Mouse
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.