missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PDK-1, His Tag Recombinant Protein

Artikelnummer. 15478393
Change view
Click to view available options
Menge:
1 mg
100 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
Artikelnummer. Menge unitSize
15478393 100 μg 100 Mikrogramm
10104392 1 mg 1 Milligramm
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 15478393

Marke: Invitrogen™ PR4194C

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

PDK1 (pyruvate dehydrogenase kinase, isozyme 1) gene, located on chromosome 2q31.1, encodes a member of the pyruvate dehydrogenase kinase family. This kinase plays a crucial role in the regulation of glucose metabolism by phosphorylating and inactivating the pyruvate dehydrogenase complex (PDC), which converts pyruvate to acetyl-CoA, linking glycolysis to the Krebs cycle. Structurally, the PDK1 protein contains a kinase domain that is essential for its enzymatic activity. PDK1 acts as a gatekeeper of glucose oxidation and is tightly regulated by various metabolic signals, including insulin and hypoxia. Dysregulation of PDK1 activity has been implicated in metabolic disorders such as diabetes and cancer, where it contributes to altered cellular energy homeostasis and metabolic reprogramming.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q15118
Konzentration See Label
Zur Verwendung mit (Anwendung) Kinase Assay
Zusammensetzung 50mM tris with 0.05% Triton X-100, 0.5mM EGTA, 20% glycerol, 100mM NaCl, 0.5mM EDTA, 5mM DTT and no preservative; pH 7.5
Gen-ID (Entrez) 5163
Molekulargewicht 66.9 kDa
Name Human PDK-1, His Tag
pH-Bereich 7.5
Reinigungsverfahren Purified
Menge 100 μg
Lagerungsbedingungen -80°C, Avoid Freeze/Thaw Cycles
Kennzeichnung RUO
Gen-Alias [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 1, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 1, mitochondrial; B830012B01; D530020C15Rik; isoenzyme 1; kinase isoenzyme 1; lipoamide; mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1; PDH kinase 1; PDHK1; PDK p48; Pdk1; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 1; Pyruvate dehydrogenase kinase isoform 1; pyruvate dehydrogenase kinase, isoenzyme 1; pyruvate dehydrogenase kinase, isozyme 1
Gebräuchliche Bezeichnung PDK1
Gensymbol PDK1
Kinase-Familie PDHK Kinase Family
Produkttyp Protein
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung His-tag
Sequenz Full length
Inhalt und Lagerung -80°C, Avoid Freeze/Thaw Cycles
Kinase-Gruppe Ser⁄Thr Kinases: Atypical
Versandbedingung Dry Ice
Expressionssystem Baculovirus
Proteinfamilie Kinases & Inhibitors
Proteinform Recombinant, Full Length
Form Liquid
Protein-Subtyp Serine/Threonine Kinases
Forschungskategorie Signal Transduction
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.