missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPD Antibody (OTI5D5), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-44638
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PEPD Monoclonal antibody specifically detects PEPD in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot
Spezifikation
| PEPD | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI5D5 | |
| Western Blot : 1:200 - 1:1000, Flow Cytometry 1:100 | |
| aminoacyl-L-proline hydrolase, EC 3.4.13.9, Imidodipeptidase, MGC10905, peptidase Dimidodipeptidase, PRD, PROLIDASE, Proline dipeptidase, xaa-Pro dipeptidase, X-Pro dipeptidase | |
| Full length human recombinant protein of human PEPD (NP_000276) produced in HEK293T cell. | |
| 100 μg | |
| metabolism | |
| 5184 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur