Learn More
Gibco™ PeproGMP® Human EGF Recombinant Protein, PeproTech®
Beschreibung
Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.
Spezifikation
Spezifikation
| Zugriffsnummer | P01133 |
| Zur Verwendung mit (Anwendung) | Control, Western Blot Control, Bioactivity |
| Format | Lyophilized |
| Zusammensetzung | Protein with no preservative |
| Gen-ID (Entrez) | 1950 |
| Molekulargewicht | 6.2 kDa |
| Name | PeproGMP™ Human EGF |
| Reinigungsverfahren | Purified |
| Menge | 100 μg |
| Lagerungsbedingungen | -20°C or -80°C if preferred |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.