Beschreibung
Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.
Spezifikation
Spezifikation
| Klassifikation | Polyclonal |
| Verdünnung | Western Blot 2 ug/ml, Simple Western 1:50, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 35041644), Dual RNAscope ISH-IHC |
| Gensymbole | PLIN2 |
| Reinigungsverfahren | Affinity Purified |
| Inhalt und Lagerung | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |