Beschreibung
Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.
Spezifikation
Spezifikation
| Klassifikation | Polyclonal |
| Zielspezies | Human, Mouse |
| Isotype | IgG |