missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripherin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Marke: Novus Biologicals NB300-137-0.025ML
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Peripherin Polyclonal specifically detects Peripherin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spezifikation
| Peripherin | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| Rabbit | |
| 57 kDa | |
| 0.025 mL | |
| Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 5630 | |
| Pig, Bovine, Rat, Human, Mouse | |
| Antisera |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Whole antisera with 5mM Sodium Azide | |
| PRPH | |
| Full length recombinant human peripherin expressed in and purified from E. coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Specifically recognizes peripherin, showing no cross reactivity with other Class III intermediate filament proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur