Få mere at vide
Få mere at vide
Peripherin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB300-137
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Peripherin Polyclonal specifically detects Peripherin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Tekniske data
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 57 kDa | |
| Human, Mouse, Pig, Bovine |
| Supplied as serum with 5mM Sodium Azide | |
| Unpurified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion