Learn More
3M™ Petrifilm Platten zur Bestimmung der Enterobakterienzahl
Product Code. 10471245 Shop All 3M Products
Change view
Click to view available options
Quantity: 50 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10471245 Quantity: 50 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10471245 Supplier 3M™ Supplier No. 7100039425
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Specifications
Produkttyp Count Plate
Zur Verwendung mit (Anwendung) Identifies contamination in the environment and food samples in as few as 22 hours.
Verpackungsart 50 Pk.
Probentyp Salmonella, Shigella and Yersini
Spezies Salmonella species, Shigella species, Yersinia species