missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PFKM Polyclonal Antibody

Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
Spezifikation
Spezifikation
| Antigen | PFKM |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.2 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with trehalose and no preservative |
| Gen | PFKM |
| Gen-Zugriffsnummer | P08237, P47857, P47858 |
| Gen-Alias | 6-phosphofructo-1-kinase; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; AI131669; ATP-dependent 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type {ECO:0000255; ATP-PFK; ATP-PFK {ECO:0000255; GSD7; HAMAP-Rule:MF_03184}; PFK; PFK1; PFK-1; Pfk4; Pfk-4; Pfka; PFK-A; PFKM; Pfk-m; PFKX; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase 4; phosphofructokinase M variant 3/4; phosphofructokinase muscle; phosphofructokinase, muscle; phosphofructokinase, polypeptide X; phosphofructokinase-1 A isozyme; phosphofructokinase-A; phosphofructokinase-M; phosphofructokinase-M (connecting peptide); phosphohexokinase; phosphohexokinase {ECO:0000255; PPP1R122; protein phosphatase 1, regulatory subunit 122 |
| Gensymbole | PFKM |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?