missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGC1 alpha Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP1-04676PE
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PGC1 alpha Polyclonal antibody specifically detects PGC1 alpha in Human, Mouse, Rat, Porcine, Goat, Hamster, Sheep, Squirrel samples. It is validated for Flow Cytometry, Flow (Intracellular), KnockDown
Spezifikation
| PGC1 alpha | |
| Polyclonal | |
| Flow Cytometry, Flow (Intracellular), Knockout Validated | |
| LEM6, Ligand effect modulator 6, ligand effect modulator-6, L-PGC-1alpha, peroxisome proliferative activated receptor, gamma, coactivator 1, alpha, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT, peroxisome proliferator-activated receptor gamma coactivator 1-alpha, peroxisome proliferator-activated receptor gamma, coactivator 1 alpha, PGC1, PGC-1(alpha), PGC1A, PGC-1-alpha, PGC1APGC-1(alpha), PGC1peroxisome proliferative activated receptor, gamma, coactivator 1, PGC-1v, PPAR gamma coactivator variant form, PPAR gamma coactivator-1, PPARgamma coactivator 1alpha, PPAR-gamma coactivator 1-alpha, PPARGC1, PPARGC-1-alpha | |
| This PGC1 alpha Antibody was developed against a recombinant protein made to an internal portion of the human PGC-1 alpha protein (within residues 400-550). [Swiss-Prot# Q9UBK2]. | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Epigenetics, Lipid and Metabolism, Mitochondrial Mediated Pathway, mTOR Pathway, Neuroscience, Transcription Factors and Regulators | |
| 10891 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, KnockDown | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Goat, Hamster, Sheep, Squirrel | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction