Learn More
Invitrogen™ PGD Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marke:  Invitrogen™ PA583189
Artikelnummer. 16380145
  • 524.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Immunogen sequence: MPGGNKEAWP HIKTIFQGIA AKVGTGEPCC DWVGDEGAGH FVKMVHNGIE YGDMQLICEA YHLMKDVLGM AQDEMAQAFE DWNKTELDSF LIEI Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 67%.
6-phosphogluconate dehydrogenase is the second dehydrogenase in the pentose phosphate shunt. Deficiency of this enzyme is generally asymptomatic, and the inheritance of this disorder is autosomal dominant. Hemolysis results from combined deficiency of 6-phosphogluconate dehydrogenase and 6-phosphogluconolactonase suggesting a synergism of the two enzymopathies.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
PGD
Polyclonal
Unconjugated
PGD
0610042A05Rik; 6PGD; 6-phosphogluconate dehydrogenase, decarboxylating; 6-phosphogluconate dehydrogenase, decarboxylating-like; AU019875; C78335; LOC100360180; Pgd; PGDH; phosphogluconate dehydrogenase
Rabbit
Antigen affinity chromatography
RUO
5226
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P52209
PGD
Recombinant protein corresponding to Human PGD. Recombinant protein control fragment (Product #RP-104627).