missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGM1 Recombinant Rabbit Monoclonal Antibody (24GB4115)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557158
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PGM1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Phosphoglucomutases (PGM; EC 5.4.2.2) catalyze the transfer of phosphate between the 1 and 6 positions of glucose. Isozymes of PGM are monomeric, with molecular masses of about 60 kD, and are encoded by several genes, including PGM1. In most cell types, PGM1 isozymes predominate, representing about 90% of total PGM activity. One exception is red cells, where PGM2 (MIM 172000) is a major isozyme (Putt et al., 1993 [PubMed 8257433]). [supplied by OMIM].
Spezifikation
| PGM1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PGM1 | |
| 3230402E02Rik; Aciculin; CDG1T; Glucose phosphomutase 1; glucose phosphomutase 2; GSD14; PGM; PGM 1; PGM 2; Pgm1; Pgm-1; Pgm2; PGM5; PGMRP; PGM-RP; Phosphodeoxyribomutase; phosphoglucomutase 1; phosphoglucomutase 2; phosphoglucomutase 5; phosphoglucomutase isoform 2; phosphoglucomutase isoform1; phosphoglucomutase-1; Phosphoglucomutase-2; phosphoglucomutase-like protein 5; Phosphoglucomutase-related protein; phosphopentomutase; zgc:63718 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24645, 5236, 66681 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4115 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36871, P38652, Q9D0F9 | |
| PGM1 | |
| A synthesized peptide derived from human PGM1 (aa 530-562). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur