Learn More
PGP9.5 / UchL1 (pan-Neuronal Marker) Mouse, Clone: rUCHL1/775, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 7345-MSM5-P0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| PGP9.5 / UchL1 (pan-Neuronal Marker) | |
| Cytoplasmic. Endoplasmic Reticulum membrane | |
| rUCHL1/775 | |
| Unconjugated | |
| Mouse | |
| 20-30kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 7345 | |
| IgG1 κ |
| Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human UCHL1 protein | |
| Cerebellum | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |