missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenforminhydrochlorid, Mikromol™
Beschreibung
Phenforminhydrochlorid
Spezifikation
Spezifikation
| Chemischer Name oder Material | Phenformin Hydrochloride |
| CAS | 834-28-6 |
| Bezeichnung der Analyten oder Bestandteile | Phenformin Hydrochloride |
| Versandbedingung | Room Temperature |
| Summenformel | C10 H15 N5 . Cl H |
| Zertifizierung/Konformität | ISO 17034 |
| Synonym | Imidodicarbonimidic diamide, N-(2-phenylethyl)-, hydrochloride (1:1), Biguanide, 1-phenethyl-, monohydrochloride (8CI), Imidodicarbonimidic diamide, N-(2-phenylethyl)-, monohydrochloride (9CI), 1-(2-Phenylethyl)biguanide hydrochloride, 1-Phenethylbiguanide hydrochloride, Azucaps, DBI-TD, Debeone DT, Debinyl, Dibein, Dipar, Feguanide, Glucopostin, Insoral, Lentobetic, Meltrol, N-Phenethylbiguanide hydrochloride, Normoglucina, Phenethylbiguanide hydrochloride, Phenformin hydrochloride, N'-beta-Phenylethylbiguanide-hydrochloride, alpha-Phenethylbiguanide hydrochloride, 1-beta-Phenylethylbiguanide hydrochloride |
| InChI-Formel | InChI=1S/C10H15N5.ClH/c11-9(12)15-10(13)14-7-6-8-4-2-1-3-5-8;/h1-5H,6-7H2,(H6,11,12,13,14,15);1H |
| SMILES | Cl.NC(=N)NC(=N)NCCc1ccccc1 |
| IUPAC-Name | 1-Carbaminidoyl-3-(2-phenylethyl)guanidin;hydrochlorid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?